missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3 Å, 8 to 12mesh, Honeywell Fluka™

Beads, 8-12 mesh

$650.13 - $1,898.68

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
6002073
View Documents Promotion Details
Honeywell-Fluka
2085821KG
1 kg
Each for $650.13
Only null left
 
6002074
View Documents Promotion Details
Honeywell-Fluka
2085825KG
5 kg
Each for $1,898.68
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

308080-99-1
202.135
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
AlNaO6Si2
MFCD00131613
19758701
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

8 to 12mesh
AlNaO6Si2
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
19758701
308080-99-1
KnNa12-n[(AlO2)12(SiO2)12] · xH2O
NONH for all modes of transport
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
202.135
Molecular Sieves, 3 Å
Videos
SDS
Documents

Documents