missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3 Å, 4 to 8mesh, Solstice

Catalog No. 6002072
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002072 5 kg
6002071 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002072 Supplier Solstice Supplier No. 2085745KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beads, 4-8 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Quantity 5 kg
Mesh Size 4 to 8mesh
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular Sieves, 3 Å