missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 13X, Pellets, Dia.: 3.2mm, Solstice

Catalog No. 60046254
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046254 1 kg Poly Bottle
60046255 5 kg Drum with inliner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046254 Supplier Solstice Supplier No. 3343591KG

Pellets, 3.2 mm

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 1 kg
Packaging Poly Bottle
Particle Size Dia.: 3.2mm
Physical Form Pellets
Linear Formula Na86[AlO2)86(SiO2)106] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular sieves, 13X
Hazard Statement P261-P305 + P351 + P338