missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 13X, Pellets, Dia.: 1.6mm, Honeywell Fluka™
Pellets, 1.6 mm diameter
$640.87 - $640.87
Chemical Identifiers
| CAS | 63231-69-6 |
|---|---|
| MDL Number | MFCD00131613 |
| IUPAC Name | Molecular sieves 13X |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 63231-69-6 | |
| MFCD00131613 | |
| Molecular sieves 13X |
Specifications
| Poly Bottle | |
| Dia.: 1.6mm | |
| 63231-69-6 | |
| MFCD00131613 | |
| Molecular sieves 13X |
| 1 kg | |
| Pellets | |
| Na86[AlO2)86(SiO2)106] · xH2O | |
| NONH for all modes of transport | |
| Molecular sieves, 13X |
Safety and Handling
P261-P305 + P351 + P338