missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 13X, 8 to 12 mesh

CAS: 63231-69-6

$143.66 - $647.20

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC197315000
SDS View Documents
Thermo Scientific Chemicals
197315000
500 g Glass bottle
Each for $143.66
Only null left
 
AC197310050
SDS View Documents
Thermo Scientific Chemicals
197310050
5 kg Plastic drum
Each for $647.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
Specifications

Chemical Identifiers

63231-69-6
MFCD00131613
Molecular sieves 13X

Specifications

23.5% min.
500 g
1.5% max.
Beads
MFCD00131613
Molecular sieves 13X, 13X, 8 to 12 mesh
Beige
Glass bottle
600g/L min.
63231-69-6
Molecular sieves 13X
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

SDS
Documents

Documents

RUO – Research Use Only