missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads

Catalog No. AAL0608530
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0608530 250 g
AAL060850B 1000 g
AAL060850I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0608530 Supplier Thermo Scientific Chemicals Supplier No. L0608530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63231-69-6

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves, 13X, 3 to 5mm (0.12 to 0.20 in) beads
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only