missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 3-4mm (0.12-0.16in) dia. pellets

Catalog No. AA33550A1
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33550A1 1 kg
AA33550A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33550A1 Supplier Thermo Scientific Chemicals Supplier No. 033550A1

CAS: 63231-69-6

Some of the major applications include commercial gas drying, air plantfeed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 1 kg
Particle Size 1/8 in.
Sensitivity Hygroscopic
Physical Form 1/8in Pellets
Solubility Information It is insoluble in water.
Chemical Name or Material Molecular sieves 13X, 3 to 4mm (0.12 to 0.16 in.) dia. pellets
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only