missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 1-2mm (0.04-0.08in) dia. pellets

Catalog No. AA87954A1
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA87954A1 1 kg
AA87954A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA87954A1 Supplier Thermo Scientific Chemicals Supplier No. 087954A1

CAS: 63231-69-6

Molecular sieves is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is preferred for static dehydration in closed liquid or gas systems, e.g., in packaging of drugs, electric components and perishable chemicals; water scavenging in printing and plastics systems and drying saturated hydrocarbon streams. Generally considered a universal drying agent in polar and nonpolar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 1 kg
Sensitivity Hygroscopic
Particle Size 1/16 in.
Physical Form 1/16in Pellets
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves, 13X, 1 to 2mm (0.04 to 0.08 in.) dia. pellets
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only