missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieve Dehydrate Fluka with indicator for drying solvents, Honeywell Fluka™
BEADS, WITH INDICATOR FOR DRYING SOLVENTS
$471.29 - $1,703.50
Specifications
| Chemical Name or Material | Molecular Sieve Dehydrate Fluka with indicator for drying solvents |
|---|---|
| CAS | 7646-79-9 , 1318-02-1 |
| MDL Number | MFCD00147630 |
| UN Number | UN3077 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Molecular Sieve Dehydrate Fluka with indicator for drying solvents | |
| 7646-79-9, 1318-02-1 | |
| MFCD00147630 | |
| UN3077 |
Safety and Handling
P201-P261-P280-P284-P305 + P351 + P338 + P310-P308 + P313