missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zymosan A (S. cerevisiae) BioParticles™, Alexa Fluor™ 594 conjugate

Catalog No. z23374
Encompass
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z23374 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z23374 Supplier Invitrogen™ Supplier No. Z23374

Study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry

  • Fluorescence of fluorescein-labeled BioParticles conjugates, which is partially quenched in acidic environments, fluorescence of the Alexa Fluor, BODIPY™ FL, tetramethylrhodamine and Texas Red™ dye conjugates is uniformly intense over the pH range from 3 to 10
  • Label (Ex/Em): Alexa Fluor594 (approx. 590/617nm)
  • Particle: Zymosan (S. cerevisiae)
  • Provided as lyophilized powders
  • Approximately 3 x 108 E. coli or S. aureus particles/mg solid and approximately 2 x 107 zymosan particles/mg solid
  • Conjugates can be reconstituted in buffer of choice for use in phagocytosis assays
  • Fluorescence of BioParticles conjugates that are bound to surface of cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
  • Addition to cellular applications, fluorescent BioParticles conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
  • Sall particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Supplement
Dye Type Alexa Fluor™ 594
Form Lyophilized Powder
pH 3 to 10
Product Line Alexa Fluor, BioParticles
Quantity 2 mg
Shipping Condition Room Temperature
Species S. cerevisiae
Show More Show Less
Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

What can the BioParticles product line be used for?

Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.

For Research Use Only. Not for use in diagnostic procedures.