missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViaGram™ Red+ Bacterial Gram Stain and Viability Kit

Catalog No. V7023
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V7023 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V7023 Supplier Invitrogen™ Supplier No. V7023
  • ONLINE EXCLUSIVE PRICE
    $469.00
    $384.58 / Each

    Save $84.42

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simple fluorescent staining protocol for fluorescence microscopy

The ViaGram Red+ Bacterial Gram Stain and Viability Kit differentially stains many gram-positive and gram-negative bacterial species and, at the same time, discriminates live from dead cells on the basis of plasma membrane integrity.

Analysis of Bacteria, Bacterial Viability & Cytotoxicity Analysis, Cell Analysis, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Cellular Imaging, Gram Staining, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbiological Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 Kit
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Conjugate Other Labels or Dyes
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Format Liquid

For Research Use Only. Not for use in diagnostic procedures.