missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Transferrin From Human Serum, Alexa Fluor™ 594 Conjugate

Catalog No. T13343
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13343 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13343 Supplier Invitrogen™ Supplier No. T13343

Labeled LDL complexes useful in studying the delivery of up to two Fe3+ atoms via receptor-mediated endocytosis

Transferrin is a monomeric serum glycoprotein (approx. 80,000 daltons) that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis.

  • Once iron-carrying transferrin proteins are inside endosomes, acidic environment favors dissociation of iron from transferrin-receptor complex
  • Following release of iron, apotransferrin is recycled to plasma membrane, where it is released from its receptor to scavenge more iron
  • Fluorescent transferrin conjugates can therefore be used with fluorescent LDL to distinguish lysosomally directed and recycling endosomal pathways
  • Experiments are typically performed by adding fluorescently labeled transferrin to cultured cells and analyzing them by microscopy
  • Label (Ex/Em): Alexa Fluor 594 (590/617)
  • Amount: 15mg solid (contains 5mg of transferrin conjugate)
  • Applications include: Image transferrin receptor dynamics using FRET; Observe receptor trafficking in live cells by confocal laser-scanning microscopy; Investigate events occurring during endosomal acidification; Measure transferrin receptor binding affinity in mammals and parasites

Cell Analysis, Cell Structure, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Transferrin
Dye Type Alexa Fluor Dyes
Form Solid
pH 7.2
Product Line Alexa Fluor
Quantity 5 mg
Shipping Condition Room Temperature
Excitation/Emission 590/617
Protein Family Transferrin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.