missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetramethylrhodamine, Methyl Ester, Perchlorate (TMRM)

Catalog No. T668
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T668 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T668 Supplier Invitrogen™ Supplier No. T668

For use with fluorescence microscopy

Tetramethylrhodamine, methyl ester (TMRM) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.

Apoptosis, Cell Analysis, Cell Structure, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Mitochondria, Mitochondria Function, Mitochondrial Function

Order Info

Shipping Condition: Room Temperature

Specifications

Color Red-Orange
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type TMRM
Quantity 25 mg
Shipping Condition Room Temperature
Sub Cellular Localization Mitochondria
I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?

If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).

What is the difference between fast and slow-response membrane potential probes?

Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.

What type of membrane potential indicators do you offer and how should I choose one for my experiment?

A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).

I am testing mitochondrial membrane potential, but my untreated cells are fluorescing, and I'm not seeing a significant difference in my test sample.

Regardless of which dye you use - tetramethylrhodamine, methyl ester (TMRM), JC-1 or MitoTracker - untreated cells will fluoresce. It's just that cells with reduced mitochondrial membrane potential will fluoresce less. It is the degree of change which is important. JC-1 dye not only changes intensity, but has a ratiometric spectral change in excitation and emission. It is very important to have an untreated control as well as a positive control treated with a mitochondrial membrane potential destabilizer, such as CCCP or FCCP. Most mitochondrial stains are only for use with live cells, as the signal will not be retained to the same degree with fixation.

For Research Use Only. Not for use in diagnostic procedures.