missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetramethylrhodamine, Ethyl Ester, Perchlorate (TMRE)

Catalog No. t669
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T669 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T669 Supplier Invitrogen™ Supplier No. T669

Tetramethylrhodamine, ethyl ester (TMRE) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.

Tetramethylrhodamine, ethyl ester (TMRE) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.

Dissolve the dye in high-quality anhydrous DMSO or ethanol to prepare stock concentration at 1 to 10 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Red-Orange
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type TMRE
Quantity 25 mg
Shipping Condition Room Temperature
Sub Cellular Localization Mitochondria
Excitation/Emission 549/574 nm
Molecular Weight (g/mol) 514.96
I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?

If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).

What is the difference between fast and slow-response membrane potential probes?

Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.

What type of membrane potential indicators do you offer and how should I choose one for my experiment?

A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).

For Research Use Only. Not for use in diagnostic procedures.