missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ 17 Red Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO

Catalog No. S7579
Encompass
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7579 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7579 Supplier Invitrogen™ Supplier No. S7579
  • ONLINE EXCLUSIVE PRICE
    $479.00
    $392.78 / Each

    Save $86.22

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with fluorescence microscopy

The cell-permeant SYTO 17 red fluorescent nucleic acid stain exhibits bright, red fluorescence upon binding to nucleic acids. Because the staining pattern of the SYTO dyes in live cells may vary between cell types, we offer the SYTO Red Fluorescent Nucleic Acid Stain Sampler Kit (Cat. No. S-11340) to enable researchers to find the most appropriate red-fluorescent SYTO stain for their system.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Red
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Excitation Wavelength Range 621 nm
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 250 μL
Volume (Metric) 250 μL
Detection Method Fluorescence
Description SYTO™ 17 Red Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Emission 634 nm
Product Line SYTO
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleic Acids
Show More Show Less
How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

For Research Use Only. Not for use in diagnostic procedures.