missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sulforhodamine 101

Catalog No. S359
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S359 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S359 Supplier Invitrogen™ Supplier No. S359

Water-soluble, nonfixable red fluorescent (excitation/emission maxima approximately 586/605nm) polar tracer

  • Can be used for investigating neuronal morphology, preparing fluorescent liposomes, studying cell-cell communications and following phagosome-lysosome uptake and fusion

Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 25 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Product Type Sulforhodamine

For Research Use Only. Not for use in diagnostic procedures.