missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SP-DiOC18(3) (3,3'-Dioctadecyl-5,5'-Di(4-Sulfophenyl)Oxacarbocyanine, Sodium Salt)

Catalog No. D7778
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D7778 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D7778 Supplier Invitrogen™ Supplier No. D7778

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

When applied to cells, dye diffuses laterally within the plasma membrane

The green fluorescent, lipophilic carbocyanine SP-DiOC18(3) is a DiO analog that contains sulfonate groups to improve water solubility. It is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. The sulfonate groups incorporated into this DiI analog improves water solubility. It has an extremely high extinction coefficient and short excited-state lifetimes (∽1 nanosecond) in lipid environments.

Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

Specifications

Color Green
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 5 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Liphophilic Tracer
Sub Cellular Localization Plasma Membrane
I stained my cells with a lipophilic cyanine dye, like DiI, but the signal was lost when I tried to follow up with antibody labeling. Why?

Since these dyes insert into lipid membranes, any disruption of the membranes leads to loss of the dye. This includes permeabilization with detergents like Triton X-100 or organic solvents like methanol. Permeabilization is necessary for intracellular antibody labeling, leading to loss of the dye. Instead, a reactive dye such as CFDA SE should be used to allow for covalent attachment to cellular components, thus providing for better retention upon fixation and permeabilization.

How long does it take for lipophlic tracers to transport along the membrane? How much faster are the FAST lipophilic dyes?

The transport is fairly slow, around 6 mm/day in live tissue and slower in fixed tissue, so diffusion of lipophilic carbocyanine tracers from the point of their application to the terminus of a neuron can take several days to weeks The FAST DiO and DiI analogs (which have unsaturated alkyl tails) can improve transport rate by around 50%.

Which form of the lipophilic tracers (DiO, DiI, DiD, etc) should I use?

Select the dye that is compatible with your available excitation source(s) and emission filter set/channels. The solid, paste and crystal forms can be applied directly to neurons in tissues. For labeling cells in culture or microinjection, the lipophilic dyes in solution or solid form can be used.

I want to label two cell populations and then perform a cell fusion assay. Which reagents are best for imaging this?

Lipophilic cyanine dyes are preferred for this sort of assay, since they insert into cellular membranes and then, upon fusion, are shared by the fused cells as the membranes are shared. For example, one cell population can be labeled with DiI (orange-red) and another cell population can be labeled with DiO (green), and when the cells fuse, the combined color appears yellow (when imaged with a dual-bandpass filter set).

For Research Use Only. Not for use in diagnostic procedures.