missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sodium Green™, Tetra (Tetramethylammonium) Salt, cell impermeant

Catalog No. S6900
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S6900 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S6900 Supplier Invitrogen™ Supplier No. S6900

Visible light-excitable Na+ indicator

  • For use with fluorescence microscope and flow cytometer
  • Fluorescent dye-based with sodium indicator

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Sodium Indicator
Dye Type Sodium Indicator
Product Line Sodium Green
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.