Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Stain
Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein-specific antibodies.
$262.00 - $3,605.00
Specifications
| Content And Storage | Store at room temperature and protect from light. |
|---|---|
| Description | Diamond Phosphoprotein Gel Stain |
| Detection Location | In-Gel Detection |
| Detection Method | Fluorescence |
| Label or Dye | Pro-Q Diamond |
Description
Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein-specific antibodies. It is ideal for the identification of kinase targets in signal transduction pathways and for phosphoproteomic studies.
Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein specific antibodies. It is ideal for the identification of kinase targets in signal transduction pathways and for phosphoproteomic studies.Features include:
• Sensitive—detection level down to 1–16 ng of phosphoprotein per band
• Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein specific antibodies and western blot analysis
• Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitative western blotting
Order Info
Shipping Condition: Room Temperature
Specifications
| Store at room temperature and protect from light. | |
| In-Gel Detection | |
| Pro-Q Diamond | |
| Proteins (Phosphoproteins) | |
| Phosphoprotein Gel Stain |
| Diamond Phosphoprotein Gel Stain | |
| Fluorescence | |
| Room Temperature | |
| PRO-Q |