missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Stain

Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein-specific antibodies.

Supplier:  Invitrogen™ P33300

Encompass

 View more versions of this product

Catalog No. P33300


Only null left

Description

Description

Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein-specific antibodies. It is ideal for the identification of kinase targets in signal transduction pathways and for phosphoproteomic studies.

Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein specific antibodies. It is ideal for the identification of kinase targets in signal transduction pathways and for phosphoproteomic studies.

Features include:
Sensitive—detection level down to 1–16 ng of phosphoprotein per band
Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein specific antibodies and western blot analysis
Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitative western blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Identifiers

Store at room temperature and protect from light.
In-Gel Detection
Pro-Q Diamond
Proteins (Phosphoproteins)
Phosphoprotein Gel Stain
Diamond Phosphoprotein Gel Stain
Fluorescence
Room Temperature
PRO-Q

Specifications

Store at room temperature and protect from light.
In-Gel Detection
Pro-Q Diamond
Room Temperature
PRO-Q
Diamond Phosphoprotein Gel Stain
Fluorescence
1 L
Proteins (Phosphoproteins)
Phosphoprotein Gel Stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions