missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phen Green™ SK, Diacetate

Catalog No. P14313
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14313 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14313 Supplier Invitrogen™ Supplier No. P14313

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Green-fluorescent heavy metal indicator can be used to detect a broad range of ions

The green-fluorescent heavy metal indicator Phen Green™ SK, diacetate can be used to detect a broad range of ions, including Cu2+, Cu+, Fe2+, Hg2+, Pb2+, Cd2+, Zn2+ and Ni2+.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Detection Method Fluorescence
For Use With (Application) Heavy Metal Indicator
For Use With (Equipment) Fluorescence Microscope, Microplate Reader
Product Type Diacetate
Dye Type Phen Green
Product Line Phen Green
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.