missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oregon Green™ 488 BAPTA-5N, Hexapotassium Salt, cell impermeant

Catalog No. o6812
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O6812 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O6812 Supplier Invitrogen™ Supplier No. O6812

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visible light-excitable, low affinity Ca2+ indicator

  • Kd for Ca2+ approximately 20μM
  • Absorption/emission maxima approximately 492/517nm

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Product Line Oregon Green
Quantity 500 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.