missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucRed™ Live 647 ReadyProbes™ Reagent

Catalog No. R37106
Encompass
Change view
Click to view available options
Quantity:
6 dropper bottle(s) kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37106 6 dropper bottle(s) kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37106 Supplier Invitrogen™ Supplier No. R37106

Includes

6 x 2.5mL dropper bottles

Cell-permeant nuclear stain that emits bright, far-red fluorescence when bound to DNA

  • Formulated as room temperature-stable solution in dropper bottle
  • Tip and drip two drops per mL to stain cells
  • Exceptionally bright far-red fluorescence well separated from blue, green, and red fluorophores
  • Rapid staining of live or fixed cells without wash steps
  • Ready-to-use liquid formulation: no need to dilute, weigh, or pipet
  • Can be used for counter-staining, DNA content measurements, cell cycle, cell replication studies, and cell counting
  • Excitation /emission of 638/686nm make it optimal for use with blue (Hoechst, DAPI, CFP), green (Alexa Fluor 488, FITC, GFP), and red (Alexa Fluor 594, Texas Red, rhodamine) fluorophores in multicolor experiments
  • May be added directly to cells in full media or buffer solutions
  • No wash, cell lysis, or RNase steps are needed
  • In most cases, 2 drops/mL and an incubation time of 15 to 30 minutes will produce bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications
  • In such cases, simply add more or fewer drops until the optimal staining intensity is obtained
  • In most cases, staining intensity increases with time if cells are not washed prior to imaging
  • Detected through far-red filters

Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry of Cellular Processes, Fluorescence In Situ Hybridization, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, In Situ Hybridization (ISH), Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

Specifications

Color Far-Red
Content And Storage 6 × 2.5 mL dropper bottles

Store at ≤ 25°C

Excitation Wavelength Range 638⁄686
Dye Type Cell-Permeant
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Flow Cytometer
Quantity 6 dropper bottle(s) kit
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line NucRed, ReadyProbes
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleus
Show More Show Less
How do I get rid of precipitates in NucRed Live 647 ReadyProbes Reagent (Cat. No. R37106)?

Briefly warm NucRed Live 647 ReadyProbes Reagent to 37 degrees C and vortex.

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

For Research Use Only. Not for use in diagnostic procedures.