missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucBlue™ Fixed Cell ReadyProbes™ Reagent (DAPI)

Catalog No. R37606
Encompass
Change view
Click to view available options
Quantity:
6 vial(s) kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37606 6 vial(s) kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37606 Supplier Invitrogen™ Supplier No. R37606
  • ONLINE EXCLUSIVE PRICE
    $244.00
    $200.08 / Each

    Save $43.92

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity form of DAPI stain in room temperature-stable solution that is provided in dropper bottle

DAPI is a commonly used nuclear counterstain for fixed cells that emits blue fluorescence when bound to DNA.

  • In fluorescence microscopy, DAPI is excited with UV light and detected through blue/cyan filter
  • DAPI is typically supplied as solid that requires weighing out and diluting several thousand-fold before use, and recommended storage is generally in freezer or refrigerator
  • This version is ready to use and can be stored next to microscope or workstation
  • Spectral properties of DAPI (4′,6-diamidino-2-phenylindole), its selectivity for dsDNA, and its increase in fluorescence when bound to DNA make it excellent fluorescent nuclear counterstain ideal for use with green (Alexa Fluor 488, FITC, GFP) and red (Alexa Fluor 594, rhodamine, Texas Red, mCherry, mKate-2) fluorophores in multicolor experiments
  • Because of its high affinity to DNA, it is also frequently used for counting cells, measuring apoptosis, sorting cells based on DNA content, and as nuclear segmentation tool in high content imaging analysis
  • In most cases 2 drops/mL and an incubation of 15 to 30 minutes will produce bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications
  • Excited by UV light at 360nm when bound to DNA, with an emission maximum at 460nm
  • It is detected through blue/cyan filter
  • As preferred blue nuclear stain in fixed cell imaging experiments, well-suited for use with antibody-based applications

Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry of Cellular Processes, Fluorescence In Situ Hybridization, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, In Situ Hybridization (ISH), Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room temperature

Specifications

Color Blue
Content And Storage 6 × 2.5 mL dropper bottles

Store at ≤ 25°C

Excitation Wavelength Range 360⁄460
Dye Type DAPI
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Quantity 6 vial(s) kit
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line Molecular Probes
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleus
Show More Show Less
My DAPI labeled samples have a strong blue background signal immediately after mounting. What can I do to fix this?

Some mounting medias can have strong blue autofluorescence. If you are seeing a high blue background, it could be coming from the mountant. Try labeling the sample and view it before (using a wet mount in buffer) and after mounting to determine if the background signal is coming from the mounting media or the sample itself.

Do you offer an alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538)?

We do not have a direct alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538). However, we do have alternative primary and secondary antibodies, as well as reagents, that can be used for trilineage differentiation. Please note that we have not internally validated the use of all these reagents together.

We recommend the following primary antibodies:

Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4 (asm-1)) (Cat. No. MA5-11547)

alpha-Fetoprotein Monoclonal Antibody (AFP3) (Cat. No. 14-6583-80)

beta-3 Tubulin Monoclonal Antibody (2G10) (Cat. No. MA1-118)

We recommend the following secondary antibodies:

Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 555 (Cat. No. A21137)

Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 594 (Cat. No. A21135)

https://www.thermofisher.com/antibody/product/Goat-anti-Mouse-IgG1-Cross-Adsorbed-Secondary-Antibody-Polyclonal/A-21121

We recommend using the following reagents:

https://www.thermofisher.com/order/catalog/product/88-8824-00

NucBlue Fixed Cell ReadyProbes Reagent (DAPI) (Cat. No. R37606)

Blocker BSA (Cat. No. 37520)

DPBS (10X), no calcium, no magnesium (Cat. No. 14200075)

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

For Research Use Only. Not for use in diagnostic procedures.