missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NP-EGTA (o-Nitrophenyl EGTA, Tetrapotassium Salt, cell impermeant

Catalog No. N6802
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N6802 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N6802 Supplier Invitrogen™ Supplier No. LSN6802

Photolabile chelator that exhibits high selectivity for Ca2+

  • Upon UV illumination, its Kd increases from 80nM to >1mm
  • Can be complexed with Ca2+ to generate caged calcium complex that will rapidly deliver Ca2+ upon photolysis

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Product Type NP-EGTA
Dye Type Chelator (Non-Fluorescent)
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.