missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nonyl Acridine Orange (Acridine Orange 10-Nonyl Bromide)

Catalog No. A1372
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1372 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1372 Supplier Invitrogen™ Supplier No. A1372

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Well retained in mitochondria making it useful probe for mitochondria during isolation and after cell fusion

The mitochondrial uptake is reported to not to depend on membrane potential, unlike many other mitochondrial probes. It is toxic at high concentrations and apparently binds to cardiolipin in all mitochondria, regardless of their energetic state. This derivative has been used to analyze mitochondria by flow cytometry, to characterize multidrug resistance and to measure changes in mitochondrial mass during apoptosis.

Apoptosis, Cell Analysis, Cell Counting & Viability, Cell Counting, Viability, & Cryopreservation, Cell Culture, Cell Structure, Cell Viability, Proliferation & Function, DNA Fragmentation, Flow Cytometry, Flow Cytometry Apoptosis Assays, Mitochondria, Mitochondria Function, Mitochondrial Function, Mitochondrial Structure

Order Info

Shipping Condition: Room Temperature

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Mitochondria Probe
Sub Cellular Localization Mitochondria

For Research Use Only. Not for use in diagnostic procedures.