missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neutral Red, high purity

Catalog No. N3246
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N3246 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N3246 Supplier Invitrogen™ Supplier No. N3246

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with microplate readers and fluorescence microscopes

The high-purity neutral red is a common lysosomal probe that stains lysosomes a fluorescent red.

Cell Analysis, Cell Structure, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Lysosomes

Order Info

Shipping Condition: Room Temperature

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope, Microplate Reader
Quantity 25 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Lysosomal Probe
Sub Cellular Localization Lysosomes

For Research Use Only. Not for use in diagnostic procedures.