missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MQAE (N-(Ethoxycarbonylmethyl)-6-Methoxyquinolinium Bromide)
Description
6-methoxyquinolinium derivative and fluorescent indicator for intracellular Cl-
- Detects ion via diffusion-limited collisional quenching
- MQAE has greater sensitivity to Cl- and higher fluorescence quantum yield than SPQ
Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Confocal Microscope, Flow Cytometer |
| Product Type | MQAE |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 100 mg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.