Learn More
Invitrogen™ Low Density Lipoprotein from Human Plasma, BODIPY™ FL complex (BODIPY™ FL LDL)
Description
Human low-density lipoprotein (LDL) is large protein complex (approx. 500,000 Da) that binds to a specific receptor on the surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis. Experiments to study LDL are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively the fluorescently labeled LDL can be injected into test animals, and the distribution of label can be analyzed after specified time period.
- Label (Ex/Em): BODIPY FL (515/520)
- Acetylated: No
- Amount: 200μL (1.0mg/mL)
- Prepared from fresh human plasma approximately every two months
- Key Applications: Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization; Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay); Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Concentration | 1 mg⁄ml |
| Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
| Detection Method | Fluorescence |
| Product Type | Supplement |
| Dye Type | BODIPY Dyes |
| Form | Liquid |
| Product Line | BODIPY |
| Quantity | 200 μL |
| Shipping Condition | Wet Ice |
Frequently Asked Questions (FAQs)
We do not recommend doing this as the oxidation process (such as oxidation with copper sulphate) may result in the degradation of the BODIPY FL dye.
The LDL in BODIPY FL LDL is not acetylated.
If the BODIPY FL LDL is applied to live cells first and then fixed, the use of methanol or acetone fixation is not recommended. Exposure to alcohols, acetone, and other organic solvents can dissociate the BODIPY FL dye from the LDL.
No. Exposure to detergents and surfactants can dissociate the BODIPY FL dye from the LDL.
The BODIPY FL dye is complexed to hydrophobic domains of the LDL; it is not covalently attached.
For Research Use Only. Not for use in diagnostic procedures.