missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipopolysaccharides from Salmonella Minnesota, Alexa Fluor™ 488 conjugate

Catalog No. L23356
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L23356 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L23356 Supplier Invitrogen™ Supplier No. L23356

Bright, green-fluorescent conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes

Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Lipopolysaccharide Conjugate
Dye Type Alexa Fluor Dyes
Product Line Alexa Fluor
Quantity 100 μg
Shipping Condition Room Temperature
Excitation/Emission 495/519

For Research Use Only. Not for use in diagnostic procedures.