missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipophilic Tracer Sampler Kit
Description
The Lipophilic Tracer Sampler kit contains 1mg samples of nine different membrane stains.
Cell Analysis, Cell Tracing & Tracking, General Cell Tracing, Microbial Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store at room temperature and protect from light. |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 1 kit |
| Detection Method | Fluorescence |
| Shipping Condition | Room Temperature |
| Product Type | Liphophilic Tracer |
| Sub Cellular Localization | Cell Membranes, Lipids, Plasma Membrane |
For Research Use Only. Not for use in diagnostic procedures.