missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS LipidTOX™ Red Neutral Lipid Stain, for cellular imaging

Catalog No. H34476
Encompass
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H34476 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H34476 Supplier Invitrogen™ Supplier No. H34476

High affinity for neutral lipid droplets

The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/or neutral lipids. HCS LipidTOX Red neutral lipid stain was developed to characterize the potentially toxic effects of compounds on lipid metabolism in mammalian cell lines. The LipidTOX neutral lipid stain has an extremely high affinity for neutral lipid droplets and can be detected by fluorescence microscopy or an HCS reader. This probe is compatible with the HCS LipidTOX phospholipidosis detection reagents. HCS LipidTOX neutral lipid stains can also be used to monitor the formation and differentiation of adipocytes, a process called adipogenesis.

Cell Analysis, Cellular Imaging, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) High Content Instrument
Quantity 1 each
Detection Method Fluorescence
Product Line LipidTOX
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Lipid Stain
Sub Cellular Localization Cell Membranes and Lipids, Lipids
What kind of filter sets can I use with HCS LipidTOX neutral lipid stains?

LipidTOX Green neutral lipid stain can be imaged with filter sets appropriate for Alexa Fluor 488 dye or fluorescein. LipidTOX Red neutral lipid stain is best imaged with filter sets appropriate for Alexa Fluor 594 dye or Texas Red dye. LipidTOX Deep Red neutral lipid stain can imaged with filter sets appropriate for Alexa Fluor 647 dye or Cy5 dye.

I want to label the plasma membrane of my cells, but there are several dyes to choose from. Which one should I use?

For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).

For Research Use Only. Not for use in diagnostic procedures.