Learn More
Invitrogen™ HCS LipidTOX™ Phospholipidosis and Steatosis Detection Kit, for high-content screening, for cellular imaging
Description
Includes
Phospholipid and neutral lipid stains, nuclear stain Hoechst 33342, propranolol and cyclosporin A (control compounds), DMSO.
HCS LipidTOX Phospholipidosis and Steatosis Detection Kit offers a complete set of reagents for performing image-based high-content screening (HCS) assays to characterize phospolipidosis and steatosis, toxic side effects on lipid metabolism that can be triggered by drugs or other compounds. The kit supplies LipidTOX Red phospholipid stain and LipidTOX Green neutral lipid stain, which can be used sequentially for the analysis of phospholipidosis and steatosis, respectively, or can be used in isolation for single-parameter analysis. Both fluorescent probes can be easily detected with fluorescence microscopes and can be quantified with stand-alone image analysis software or the built-in image analysis software of most HCS readers.
Cell Analysis, Cellular Imaging, High-Content Screening (HCS)
Order Info
Shipping Condition: Room temperature. Sufficient reagents are supplied for two 96-well plates or ten 96-well plates.
Specifications
Specifications
| Content And Storage | When stored at -20°C, desiccated, and protected from light, the kit is stable for at least 6 months. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | High Content Analysis Instrument |
| Product Type | Phospholipidosis and Steatosis Detection Kit |
| Dye Type | Other Label(s) or Dye(s) |
| Format | 96-well plate |
| Product Line | LipidTOX, Molecular Probes |
| Quantity | 1200 Assays |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
After thawing LipidTOX Red phospholipidosis detection reagent, some minute aggregates might be observed in the solution. They usually will disappear if the vial is incubated in a 37 degrees C water bath for 5 minutes. These aggregates do not affect the performance of the assay.
Any aggregates that remain after the stain is diluted in media are removed by 0.2 µm filtration before the labeling solution is added to the cells, as recommended in the protocol.
For Research Use Only. Not for use in diagnostic procedures.