missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS LipidTOX™ Green Phospholipidosis Detection Reagent (1000X), for cellular imaging

Catalog No. H34350
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H34350 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H34350 Supplier Invitrogen™ Supplier No. H34350

Easily detected by fluorescence microscopy

The HCS LipidTOX Green phospholipidosis detection reagent was developed to characterize the potentially toxic side effects of compounds on lipid metabolism in mammalian cell lines. This reagent is easily detected by fluorescence microscopy or an HCS reader equipped with filters that can detect excitation/emission maxima approx. 495/525nm. LipidTOX phospholipidosis detection reagents do not affect the normal growth of cells, and the staining observed after incubation in live cells is maintained after formaldehyde fixation. This probe is compatible with the HCS LipidTOX neutral lipid stains.

Cell Analysis, Cellular Imaging, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) High Content Analysis Instrument
Product Type Phospholipidosis Detection Reagent
Dye Type Other Label(s) or Dye(s)
Format 96-well plate
Product Line LipidTOX
Quantity Each
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.