missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS LipidTOX™ Green Neutral Lipid Stain, for cellular imaging

Catalog No. H34475
Encompass
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H34475 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H34475 Supplier Invitrogen™ Supplier No. H34475

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High affinity for neutral lipid droplets

The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/or neutral lipids. HCS LipidTOX Green neutral lipid stain was developed to characterize the potentially toxic effects of compounds on lipid metabolism in mammalian cell lines. The LipidTOX neutral lipid stain has an extremely high affinity for neutral lipid droplets and can be detected by fluorescence microscopy or an HCS reader. This probe is compatible with the HCS LipidTOX phospholipidosis detection reagents. HCS LipidTOX neutral lipid stains can also be used to monitor the formation and differentiation of adipocytes, a process called adipogenesis.

Cell Analysis, Cellular Imaging, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature

Specifications

Color Green
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) High Content Instrument
Quantity 1 each
Detection Method Fluorescence
Product Line LipidTOX
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Lipid Stain
Sub Cellular Localization Cell Membranes, Lipids

For Research Use Only. Not for use in diagnostic procedures.