missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lectin HPA From Helix pomatia (edible snail), Alexa Fluor™ 488 Conjugate

Catalog No. L11271
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L11271 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L11271 Supplier Invitrogen™ Supplier No. L11271

Suitable for use in cellular imaging and flow cytometry

Lectin Helix pomatia agglutinin (HPA) selectively binds to α-N-acetylgalactosamine residues and type A erythrocytes.

Cell Analysis, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 mg
Source Edible snail
Conjugate Alexa Fluor 488
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.