missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LC3B Antibody Kit for Autophagy DFS Item

Use in fluorescence microscopy and high content imaging

Supplier:  Invitrogen™ L10382

Catalog No. L10382


Only null left
Explore available promotions

Includes: Kit includes includes a rabbit polyclonal antibody against LC3B and chloroquine diphosphate.

Description

Description

The LC3 Antibody Kit for Autophagy includes a rabbit polyclonal antibody against LC3B that has been validated for use in fluorescence microscopy and high content imaging and analysis. Chloroquine diphosphate is also included - following treatment with this compound, normal autophagic flux is disrupted and autophagosomes accumulate as a result of the disruption in normal autophagic flux.

Apoptosis, Autophagy, Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

Kit includes includes a rabbit polyclonal antibody against LC3B and chloroquine diphosphate.
  • Store at ≤ -20°C
  • Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument
    Unlabeled
    1 kit
    Fluorescence
    Dye
    96-well plate, Slide(s)
    Dry Ice
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions

    For Research Use Only. Not for use in diagnostic procedures.