missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene)

Catalog No. D250
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D250 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D250 Supplier Invitrogen™ Supplier No. D250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

When the lipophlilic probe laurdan is incorporated into membranes, its fluorescence spectra is sensitive to the physical state of the surrounding phospholipids.

When the lipophlilic probe laurdan is incorporated into membranes, its fluorescence spectra is sensitive to the physical state of the surrounding phospholipids.

To make a concentrated stock solution of up to 20 mM, dissolve in either DMF or acetonitrile. Store frozen, desiccated and protected from light.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Green
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Cell Membranes, Lipids
Molecular Weight (g/mol) 353.55
What is the recommended solvent and concentration for Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) (Cat. No. D250) stock solution?

We recommend resuspending Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) (Cat. No. D250) in dimethylformamide (DMF) or acetonitrile at a concentration of 1 mM. The molecular weight for this product is 353.55 g/mol.

The product is stable at room temperature and should be protected from light.

For Research Use Only. Not for use in diagnostic procedures.