Learn More
Invitrogen™ Isolectin GS-IB4 From Griffonia simplicifolia, biotin-XX Conjugate
Isolated from the seeds of Griffonia simplicifolia, a tropical African legume
Supplier: Invitrogen™ I21414
Description
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4). The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The biotin-XX isolectin GS-IB4 conjugate can be used in fluorescence microscopy or flow cytometry in conjunction with fluorescently labeled streptavidin.
Cell Analysis, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
| 500 μg | |
| Biotin | |
| Biotin & Other Haptens | |
| Lyophilized |
| Africa Shrub legume | |
| Store in freezer (-5 to -30°C). | |
| Room Temperature | |
| Lectins |
For Research Use Only. Not for use in diagnostic procedures.