missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-IT™ LIVE Plasma Membrane and Nuclear Labeling Kit (Counterstains For GFP-Expressing Cells)

Catalog No. I34406
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I34406 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I34406 Supplier Invitrogen™ Supplier No. I34406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Counterstains for GFP-expressing cells

The Image-iT LIVE Plasma Membrane and Nuclear Labeling Kit provides two stains - red-fluorescent Alexa Fluor 594 wheat germ agglutinin (WGA) and blue-fluorescent Hoechst 33342 dye - for highly selective staining of the plasma membrane and nucleus, respectively, of live green-fluorescent protein (GFP)-transfected cells.

Cell Analysis, Cell Structure, Cell-Based Reporter Assays, Cellular Imaging, Enzyme & Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Nucleus, Nucleoli & Nuclear Envelope, Plasma Membrane, Proteins, Expression, Isolation & Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

Specifications

Color Red, Blue
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 1 kit
Detection Method Fluorescence
Product Line Image-iT
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Membrane and Nuclear Labeling
Sub Cellular Localization Nucleus, Plasma Membrane

For Research Use Only. Not for use in diagnostic procedures.