missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-IT™ DEAD Green™ Viability Stain

Catalog No. I10291
Encompass
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10291 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I10291 Supplier Invitrogen™ Supplier No. I10291

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The stain includes sufficient material to perform 10 plates in a 96-well plate format and 50μL well volume.

Stain becomes permeant when plasma membrane is damaged

Image-iT DEAD Green viability stain is a fast, sensitive and easy-to-use viability indicator that is also amenable to fixation and permeabilization, thus allowing for multiplexing with other biomarkers of cytotoxicity. Image-iT DEAD Green is an impermeant dye to healthy cells that becomes permeant when the plasma membrane of cells is compromised.

Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function

Order Info

Shipping Condition: Room temperature

Specifications

Color Green
Concentration 1 mM
Content And Storage The stain includes sufficient material to perform 10 plates in a 96-well plate format and 50 μL well volume.
  • Store at ≤ −20°C, desiccate and protect from light.
  • Detection Method Fluorescence
    For Use With (Application) Viability Assay
    For Use With (Equipment) Fluorescence Microscope
    Product Type Viability Stain
    Form Liquid
    Product Line Image-iT, Molecular Probes
    Quantity 1 Vial
    Shipping Condition Wet Ice
    Sub Cellular Localization Nucleus
    Emission Wavelength Range Visible
    Show More Show Less
    Do you offer a cell viability stain compatible with fixation?

    Our Image-IT DEAD Green Viability Stain (Cat. No. I10291) is amenable to fixation and permeabilization after the application of the stain. It is important to use fixatives free of alcohol or other organic solvents.

    Which cell viability kits are compatible with fixation?

    The LIVE/DEAD Fixable kits for flow cytometry analysis are compatible with fixation. These kits use amine-reactive cell-impermeant dyes that stain the cell surface of live cells and also the cytosol of dead cells-live cells are dim and dead cells are bright. Since the dye is covalently bound to the cells, it will be retained after fixation. Unfortunately, this method does not work well for imaging-based assays, as all cells are stained and it is difficult to distinguish bright dead cells from dim live cells with a microscope. Ethidium monoazide (EMA; Cat No. E1374) is a cell impermeant nucleic acid stain that can be applied to live cultures and stains only dead cells. After incubation and washing away unbound dye, the cells can be exposed to light to photoactivate EMA to crosslink to dead cell DNA. After crosslinking to dead cell DNA, the samples may be fixed and permeabilized. Image-IT DEAD Green Viability Stain (Cat. No. I10291) for imaging and high-content screening (HCS) analysis is a live-cell impermeant DNA binding dye that is compatible with fixation and permeabilization with good retention up to 48 hours. We also have a LIVE/DEAD Reduced Biohazard Cell Viability Kit (Cat. No. L7013) for imaging and flow analysis that contains two DNA binding dyes, SYTO 10 and Dead Red, that are sufficiently retained to be analyzed soon after 4% glutaraldehyde fixation.
    Note: In general, DNA-binding dyes and calcein AM are not compatible with fixation, as these dyes are not covalently bound to components of the cell and will thus slowly diffuse out of cells after fixation, gradually staining all cells as dead.

    For Research Use Only. Not for use in diagnostic procedures.