missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Escherichia coli BioParticles™ Opsonizing Reagent

Catalog No. E2870
Encompass
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E2870 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E2870 Supplier Invitrogen™ Supplier No. E2870

The Escherichia coli BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies.

The Escherichia coli BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies and specifically designed to enhance the uptake of the Escherichia coli BioParticals™ conjugates.

See User Manual for solubility instructions.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Opsonizing Reagent
Dye Type Unconjugated
Form Solid
Product Line BioParticles
Quantity 1 unit
Shipping Condition Room Temperature
Species E. coli
Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

For Research Use Only. Not for use in diagnostic procedures.