missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Histone H1 From Calf Thymus, Alexa Fluor™ 488 Conjugate

Catalog No. H13188
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13188 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13188 Supplier Invitrogen™ Supplier No. H13188

Labeled with the bright and photostable dye

  • Exhibits fluorescence with excitation/emission maxima approx. 495/520nm and can be visualized by fluorescence microscopy or flow cytometry

Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Lipopolysaccharide Conjugate
Dye Type Alexa Fluor Dyes
Product Line Alexa Fluor
Quantity 1 mg
Reagent Type Phosphate Buffered Saline
Shipping Condition Room Temperature
Protein Family Histone H1

For Research Use Only. Not for use in diagnostic procedures.