missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hexidium Iodide

Catalog No. H7593
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H7593 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H7593 Supplier Invitrogen™ Supplier No. H7593

Permeant to mammalian cells

The nucleic acid stain, hexidium iodide selectively stains almost all gram-positive bacteria in the presence of gram-negative bacteria. Hexidium iodide exhibits excitation/emission maxima of ∽518/600nm upon binding DNA. Generally, both the cytoplasm and nuclei of eukaryotic cells show staining with hexidium iodide.

Analysis of Bacteria, Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Gram Staining, Microbial Tracking, Microbiological Analysis, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room Temperature

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
Excitation Wavelength Range 518⁄600
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 5 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Cytoplasm, Cytosol, Nucleus
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.