missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS DNA Damage Kit

Catalog No. H10292
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10292 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10292 Supplier Invitrogen™ Supplier No. H10292
  • ONLINE EXCLUSIVE PRICE
    $1,042.00
    $854.44 / Each

    Save $187.56

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Kit contains sufficient reagents for performing two plates in a 96-well plate format.

Detects DNA damage and cytotoxicity

The HCS DNA Damage kit simultaneously and quantitatively measures two important cell-health parameters: DNA damage and cytotoxicity. DNA damage is detected using an antibody against phosphorylated H2AX (Ser139) which is induced in response to double-strand break (DSB) formation. To detect cytotoxicity, the Image-iT™ DEAD™ Green is an impermeant dye in healthy cells that becomes permeant when the plasma membrane of cells is compromised. Both indicators are amenable to fixation and permeabilization, thus allowing for multiplexing with other biomarkers of interest. The blue-fluorescent nuclear segmentation tool Hoechst 33342 is also included and stains DNA in both live and dead cells.

Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function

Order Info

Shipping Condition: Dry ice

Specifications

Color Orange, Blue, Green
Content And Storage The kit contains sufficient reagents for performing 2 plates in a 96-well plate format.
  • Store at ≤-20°C, desiccate and protect from light.
  • Detection Method Fluorescence
    For Use With (Equipment) High Content Analysis Instrument
    Product Type DNA Damage kit
    Dye Type Alexa Fluor™ 555, Hoechst 33258
    Format 96-well plate
    Product Line Molecular Probes
    Quantity 1 kit
    Shipping Condition Dry Ice
    What are the fluorescence excitation/emission maxima for the dyes contained in the HCS DNA Damage Kit?

    Hoechst 33342: 350/461 nm, bound to DNA
    Image-IT Dead Green viability stain: 488/515 nm
    Alexa Fluor 555 goat anti-mouse IgG: 555/565 nm

    For Research Use Only. Not for use in diagnostic procedures.