missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fura Red™, AM, cell permeant

Catalog No. p-4922886
Encompass
Change view
Click to view available options
Quantity:
10 x 50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F3020 500 μg
F3021 10 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F3020 Supplier Invitrogen™ Supplier No. F3020
Only null left

Visible light-excitable fura-2 analog that offers particular possibilities for ratiometric measurement of Ca2+ in single cells

  • Measure by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicator
  • Acetoxymethyl (AM) ester form
  • Useful for noninvasive intracellular loading

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Cell-Based Second Messenger Assays, Drug Discovery and Development, G-Protein Coupled Receptor Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Product Line Fura Red
Quantity 500 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.