missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fura dextran, Potassium Salt, 10,000 MW, Anionic

Catalog No. F3029
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F3029 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F3029 Supplier Invitrogen™ Supplier No. F3029

Derivative of ratiometric, UV light-excitable fura-2 indicator coupled to water-soluble 10,000 MW dextran

  • Useful for intracellular loading by microinjection, infusion from patch pipet or uptake induced by Influx™ pinocytic cell-loading reagent (Cat. No. I-14402)
  • Fura dextran conjugates tend to remain in cytosol without compartmentalization or leakage and are less likely to bind to cellular proteins, making them useful for long-term Ca2+ measurements

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Potassium Assay
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Fura Dextran Potassium Salt
Dye Type Fluorescent Dye-Based
Quantity 5 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.