missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoZin™-3, AM, cell permeant

Catalog No. F24195
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F24195 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F24195 Supplier Invitrogen™ Supplier No. F24195

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zn2+-selective indicator with a structure that resembles fluo-4.

The FluoZin™-3 indicator is a Zn2+-selective indicator with a structure that resembles fluo-4. FluoZin™-3 exhibits high Zn2+-binding affinity that is unperturbed by Ca2+ concentrations up to at least 1 μM. In addition, FluoZin™-3 exhibits a >50-fold increase in fluorescence in response to saturating levels of Zn2+. Although imaging applications of FluoZin™-3 are predominant, cell-based microplate assays and flow cytometry protocols have also been developed. FluoZin™-3 indicator is available in a cell-permeant and a cell-impermeant form.

Fluorescent Zinc Indicators Specifications:

  • Label (Ex/Em): FluoZin™-3 (∼494/516 nm)
  • Kd (Zn2+) (in buffer): ∼15 nM
  • Lyophilized product may be dissolved in DMSO for use
  • Product is typically loaded into cells by adding the dissolved indicator to medium containing cells.

Find Fluorescent Indicators for Zinc and Other Metal Ions

We offer a number of fluorescent indicators to determine polyvalent cation concentrations inside cells, to follow metal ion transport through ion channels, to make measurements in environmental samples, and more. Review Fluorescent Indicators for Zn2+ and Other Metal Ions—Section 19.7 in the Molecular Probes™ Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Zinc Indicator
For Use With (Equipment) Fluorescence Microscope
Product Type Zinc Indicator
Dye Type FluoZin-3
Excitation Wavelength Range 480 to 495 nm
Product Line FluoZin
Quantity 100 μg
Shipping Condition Room Temperature
Molecular Weight (g/mol) 982.85 g/mol
Emission Wavelength Range 510 to 525 nm
Excitation Wavelength 494 nm
Emission Wavelength 516 nm
Show More Show Less
Why don't I see a significant change in signal for my live-cell fluorescent indicator dye?

Regardless of the type of live-cell indicator dye (e.g., calcium indicators, pH indicator, metal ion indicators), make sure there is no serum during the loading step, which can prematurely cleave dyes with AM esters and bind dyes non-specifically. Always optimize the dye concentration and staining time with a positive control before you run your test samples, to give the best signal-to-background. Always run a positive control with a buffer containing free ions of known concentration and an ionophore to open pores to those ions (for instance, for calcium indicators like Fluo-4 AM, this would include a buffer with added calcium combined with calcimycin, or for pH indicators, buffers of different pHs combined with nigericin). Reactive oxygen indicators, such as CellROX Green or H2DCFDA would require a cellular reactive oxygen species (ROS) stimulant as a positive control, such as menadione. Finally, make sure your imaging system has a sensitive detector. Plate readers, for instance, have much lower detector efficiency over background, compared to microscopy or flow cytometry.

For Research Use Only. Not for use in diagnostic procedures.