missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein Diacetate (FDA)

Catalog No. F1303
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1303 Supplier Invitrogen™ Supplier No. F1303
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell-permeant esterase substrate that can serve as viability probe

  • Measures both enzymatic activity required to activate its fluorescence and cell-membrane integrity, intracellular retention of their fluorescent product
  • Upon hydrolysis by intracellular esterases, AM ester yields fluorescein

Cell Adhesion, Cell Analysis, Cell Proliferation, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, Target-Based ADME/Tox Assays, pH Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Product Type Fluorescein Diacetate
Dye Type Classic Dyes
Quantity 1 g
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.