missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluo-5N, Pentapotassium Salt, cell impermeant

Catalog No. f14203
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F14203 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F14203 Supplier Invitrogen™ Supplier No. F14203

Analog of fluo-4 with lower Ca2+-binding affinity, making it suitable for detecting intracellular calcium levels in 1μM to 1 mM range that would saturate response of fluo-3 and fluo-4

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+.

  • Cells may be physically loaded with cell-impermeant salt forms of these indicators using patch pipet or microinjection or Influx™ pinocytotic cell-loading reagent
  • Compatible with excitation at 488nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • BAPTA-based indicators bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
  • Perturbations to calcium measurements caused by presence of the ions can be controlled using heavy metal-selective chelator
  • Label (Ex/Em of Ca2+-bound form): Fluo-5N (494/516nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: approximately 90μM

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Quantity 500 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.